missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FES Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551568
This item is not returnable.
View return policy
Description
Immunogen sequence: DSAQAKRKYQ EASKDKDRDK AKDKYVRSLW KLFAHHNRYV LGVRAAQLHH QHHHQLLLPG LLRSLQDLHE EMACILKEIL QEYLEISSLV QDEVVAIHRE MAAAAARIQP EAEYQGFLRQ YGSAPDVPPC VTFDESLLEE GEPLEP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
FES (FPS) is a tyrosine protein kinase that is required for maintenance of cellular transformation. Its chromosomal location is linked to a specific translocation event identified in patients with acute promyelocytic leukemia, and it is also involved in normal hematopoiesis. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
| FES | |
| Polyclonal | |
| Unconjugated | |
| FES | |
| AI586313; BB137047; c-fes; EC 2.7.10.2; feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami avian sarcoma (PRCII) viral (v-fps) oncogene homolog; feline sarcoma oncogene; feline sarcoma/Fujinami avian sarcoma oncogene homolog; Fes; FES proto-oncogene, tyrosine kinase; FPS; Oncogene FES, feline sarcoma virus; p93c-fes; Proto-oncogene c-Fes; proto-oncogene c-Fps; proto-oncogene tyrosine-protein kinase Fes/Fps; RGD1564385; tyrosine kinase Fps/Fes; tyrosine-protein kinase Fes/Fps; Variant 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2242 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07332 | |
| FES | |
| Recombinant protein corresponding to Human FES. Recombinant protein control fragment (Product #RP-95943). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction