missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Fes/Fps Protein, Active

Product Code. 17360214
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17360214

Brand: Eurofins DiscoverX 14473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, recombinant full length human Fes, containing an N-terminal His6-tag, for use in Kinase Assays.

The cyclin-dependent kinases (CDKs), are a family of serine threonine protein kinases, that control the cell division cycle. In mammals, nine different CDKs (cdk 1 to cdk 9) have been identified. CDKs are regulated on various levels including binding of proteins (cyclins, inhibitors, and assembly factors), protein degradation, transcriptional control, localization, and multiple phosphorylations. This leads to timely regulation of CDK activity, allowing progression from one cell cycle phase to the next. The cdk2 promotes the G1/S trasition and progression through the S phase when bound to specific cyclins (E and A). Recent evidences indicate that CDK2 is involved in cell cycle independent functions such as DNA damage repair.

GenBank X06292

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 98 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥57%
Protein Fes/Fps, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.