missing translation for 'onlineSavingsMsg'
Learn More

Ferulic acid sodium, MedChemExpress

Product Code. 16491288
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491288 5 g 5g
16452489 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491288 Supplier Medchem Express Supplier No. HYN0060A
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ferulic acid sodium is a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.

Ferulic acid sodium is a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.

Specifications

Chemical Name or Material Ferulic acid sodium
CAS 24276-84-4
Color White
Purity Grade Notes Research
Molecular Formula C10H9NaO4
Quantity 5 g
Synonym Coniferic acid sodium
Solubility Information H2O : 100 mg/mL (462.60 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (154.18 mM; Need ultrasonic)
SMILES O=C(O[Na])/C=C/C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 216.17
Formula Weight 216.17
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.