missing translation for 'onlineSavingsMsg'
Learn More

Ferulic acid, MedChemExpress

Product Code. 16468966
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468966 100 mg 100mg
16422346 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468966 Supplier Medchem Express Supplier No. HYN0060
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ferulic acid is a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.

Ferulic acid is a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.

Specifications

Chemical Name or Material Ferulic acid
CAS 1135-24-6
Color White
Purity Grade Notes Research
Molecular Formula C10H10O4
Quantity 100 mg
Synonym Coniferic acid
Solubility Information DMSO : 100 mg/mL (514.99 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 194.18
Formula Weight 194.18
Percent Purity 98.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.