missing translation for 'onlineSavingsMsg'
Learn More

Ferulic Acid-d3, TRC

Product Code. 30393096
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30393096 Supplier Toronto Research Chemicals Supplier No. TRCF308902100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-Ferulic Acid-d3
CAS 860605-59-0
Purity Grade Notes HPLC
Molecular Formula C10 D3 H7 O4
Quantity 100 mg
Synonym 2-Propenoic acid, 3-[4-hydroxy-3-(methoxy-d3)phenyl]-, (2E)- (9CI), (2E)-3-(4-Hydroxy-3-[methoxy-d3)phenyl]-2-propenoic acid, (2E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenoic acid-d3, (E)-Ferulic-D3 acid (methyl-D3), trans-4-Hydroxy-3-methoxycinnamic acid D3, trans-Ferulic-d3 acid
InChI Formula InChI=1S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+/i1D3
SMILES [2H]C([2H])([2H])Oc1cc(\C=C\C(=O)O)ccc1O
IUPAC Name (E)-3-[4-hydroxy-3-(trideuteriomethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol) 197.202
Formula Weight 197.077
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.