missing translation for 'onlineSavingsMsg'
Learn More

Ferulic Acid-d3 4-O-Sulfate Disodium Salt, TRC

Product Code. 30400557
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400557 2.5 mg 2.5mg
30395157 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400557 Supplier Toronto Research Chemicals Supplier No. TRCF3089232.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10 D3 H5 Na2 O7 S
Molecular Weight (g/mol) 321.229
Synonym 3-[3-(Methoxy-d3)-4-(sulfooxy)phenyl]-2-propenoic Acid Disodium Salt
IUPAC Name [4-[(E)-3-oxo-3-sodiooxyprop-1-enyl]-2-(trideuteriomethoxy)phenoxy]sulfonyloxysodium
SMILES [2H]C([2H])([2H])Oc1cc(\C=C\C(=O)O[Na])ccc1OS(=O)(=O)O[Na]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C10H10O7S.2Na/c1-16-9-6-7(3-5-10(11)12)2-4-8(9)17-18(13,14)15;;/h2-6H,1H3,(H,11,12)(H,13,14,15);;/q;2*+1/p-2/b5-3+;;/i1D3;;
Formula Weight 320.997
Chemical Name or Material Ferulic Acid-d3 4-O-Sulfate Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.