missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Ferrule for HPLC Systems

Product Code. 14757170 Shop All SCAT Europe Products
Change view
Click to view available options
Diameter (Metric) Inner:
1.6 mm
2.3 mm
3.2 mm
Unit Size:
Pack of 10
Product Code. Diameter (Metric) Inner unitSize
14757170 3.2 mm Pack of 10
14737170 1.6 mm Pack of 10
14747170 2.3 mm Pack of 10
3 options
This item is not returnable. View return policy

Product Code. 14757170

Brand: SCAT™ 107908

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.

SCAT™ fittings are available in the following versions:

  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters

Specifications

Type Ferrule
Color White
Diameter (English) Inner 0.13 in.
Diameter (Metric) Inner 3.2 mm
For Use With Capillary
Material PTFE
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.