missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate Heptahydrate, TRC

Product Code. 30012327
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012327 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012327 Supplier Toronto Research Chemicals Supplier No. TRCF30785010G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7782-63-0
Quantity 10 g
Molecular Formula Fe . O4 S . 7[H2 O]
Synonym Sulfuric acid, iron(2+) salt, heptahydrate (1:1:7), Sulfuric acid, iron(2+) salt (1:1), heptahydrate (8CI,9CI), FerroHepta, Ferrous sulfate heptahydrate, Iron protosulfate, Iron sulfate (FeSO4) heptahydrate, Iron sulfate heptahydrate, Iron sulfate heptahydrate (FeSO4.7H2O), Iron(2+) sulfate heptahydrate, Iron(II) sulfate heptahydrate, Ironate, Ironfer, Irosul, Presfersul
InChI Formula InChI=1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2
SMILES O.O.O.O.O.O.O.[Fe+2].[O-]S(=O)(=O)[O-]
IUPAC Name iron(2+);sulfate;heptahydrate
Molecular Weight (g/mol) 278.015
Formula Weight 277.961
Chemical Name or Material Iron(II) Sulfate Heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.