missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrostatin-1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
Ferrostatin-1 (Fer-1), a potent and selective ferroptosis inhibitor, suppresses Erastin-induced ferroptosis in HT-1080 cells (EC50=60 nM). Ferrostatin-1, a synthetic antioxidant, acts via a reductive mechanism to prevent damage to membrane lipids and thereby inhibits cell death. Antifungal Activity.
Specifications
Specifications
| Chemical Name or Material | Ferrostatin-1 |
| Physical Form | Powder |
| CAS | 347174-05-4 |
| Color | Pale Purple |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H22N2O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | Fer-1 |
| Solubility Information | DMSO : 125 mg/mL (476.46 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction