missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ferrostatin 1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ferrostatin 1 is a selective inhibitor of erastin induced ferroptosis (EC50 = 60 nM). Specifically inhibits Ras selective lethal compound -induced death, but not cell death induced by other oxidative lethal compounds and apoptosis-inducing agents. Inhibits ferroptosis in cancer cells; also inhibits glutamate-induced cell death in organotypic rat brain slices. Prevents erastin induced accumulation of cytosolic and lipid reactive oxygen species.
Spécification
Spécification
| Quantity | 10 mg |
| Product Type | Ferrostatin 1 |
| Purity | >98% |
| Molecular Weight (g/mol) | 262.35 |
| Inhibitors | Ferrostatin 1 |
For Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu