missing translation for 'onlineSavingsMsg'
Learn More

Ferroquine, MedChemExpress

Product Code. 16440316
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440316

Brand: Medchem Express HY19364

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ferroquine (Ferrochloroquine), a ferrocenyl analogue of Chloroquine, is an antimalarial agent. Ferroquine shows parasiticidal effect on Plasmodium by inducing oxidative stress and the subsequent destruction of the membrane.

Ferroquine (Ferrochloroquine), a ferrocenyl analogue of Chloroquine, is an antimalarial agent. Ferroquine shows parasiticidal effect on Plasmodium by inducing oxidative stress and the subsequent destruction of the membrane.

Specifications

Chemical Name or Material Ferroquine
Physical Form Powder
CAS 185055-67-8
Color Yellow
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C23H24ClFeN3
For Use With (Application) COVID-19-immunoregulation
Synonym Ferrochloroquine SSR97193
Solubility Information DMSO : 8.33 mg/mL (19.20 mM; Need ultrasonic)
SMILES ClC1=CC=C2C(NC[C-]34[Fe+2]56789%10%11([CH-]%12[CH]8=[CH]9[CH]%10=[CH]%11%12)C3(CN(C)C)=[CH]5[CH]6=[CH]47)=CC=NC2=C1
Molecular Weight (g/mol) 433.75
Formula Weight 433.75
Percent Purity 98.45%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.