missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ferroportin/SLC40A1 Antibody (LY2928057) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28537-50ug
This item is not returnable.
View return policy
Description
Ferroportin/SLC40A1 Monoclonal antibody specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Ferroportin/SLC40A1 | |
| Monoclonal | |
| Unconjugated | |
| Q9NP59 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| LY2928057 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
| SLC40A1 | |
| 50 μg | |
| Cancer, Lipid and Metabolism, Signal Transduction | |
| 30061 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction