missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ferroportin Monoclonal Antibody (C9), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £361.00
Specifications
| Antigen | Ferroportin |
|---|---|
| Clone | C9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17960553
![]() |
Invitrogen™
MA550837 |
200 μL |
£361.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17939972
|
Invitrogen™
MA550838 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.Specifications
| Ferroportin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| CAR1; Cell adhesion regulator; duodenal-specific; Dusg; Ferroportin; ferroportin 1; ferroportin-1; Fpn1; Fpt1; HFE4; IREG1; iron regulated gene 1; iron-regulated transporter 1; Metal transporter protein 1; metal transporting protein 1; MST079; MSTP079; MTP; MTP1; Ol5; Pcm; polycythaemia; Slc11a3; SLC11A3 iron transporter; Slc39a1; Slc40a1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 39 (iron-regulated transporter), member 1; solute carrier family 40 (iron-regulated transporter), member 1; solute carrier family 40 member 1 | |
| SLC40A1 | |
| IgG2b κ | |
| Protein A/G | |
| SLC40A1 |
| C9 | |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q9JHI9, Q9NP59 | |
| 30061, 53945 | |
| Recombinant Human FPN protein, Trp127-Val321 (Accession #Q9NP59). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title