missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Ferroportin Monoclonal Antibody (C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541585
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Ferroportin Monoclonal Antibody for Western Blot
SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.
Tekniske data
| Ferroportin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9NP59 | |
| SLC40A1 | |
| Recombinant protein Ferroportin. The antigen corresponds to amino acid range 127-321 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C7 | |
| Unconjugated | |
| SLC40A1 | |
| CAR1; Cell adhesion regulator; duodenal-specific; Dusg; Ferroportin; ferroportin 1; ferroportin-1; Fpn1; Fpt1; HFE4; IREG1; iron regulated gene 1; iron-regulated transporter 1; Metal transporter protein 1; metal transporting protein 1; MST079; MSTP079; MTP; MTP1; Ol5; Pcm; polycythaemia; Slc11a3; SLC11A3 iron transporter; Slc39a1; Slc40a1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 39 (iron-regulated transporter), member 1; solute carrier family 40 (iron-regulated transporter), member 1; solute carrier family 40 member 1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 30061 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion