missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ FerroOrange

Product Code. 31900351
Change view
Click to view available options
Quantity:
24 μg
Unit Size:
24µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31900351 24 μg 24µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31900351 Supplier R&D Systems™ Supplier No. 8846/24U
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Sensitive and cell-permeable fluorogenic probe for the detection of labile Fe(II) in live cells; suitable for high-throughput assays

Key information: FerroOrange (RhoNox-4) is a highly sensitive and robust fluorogenic probe for the detection of intracellular labile Fe(II). FerroOrange displays weak absorption and low fluorescence in the visible region, but becomes strongly fluorescent upon reaction with Fe(II).

Used for: detection of intracellular labile Fe(II).

Application: high-throughput assays for high content imaging of subcellular labile Fe(II) in live cells; fluorescent microscopy; studies on iron homeostasis and ferroptosis.

Properties and Photophysical Data: FerroOrange has excellent cell-membrane permeability and low cell toxicity. FerroOrange has no chelating ability. For the fluorescent form, excitation and emission maxima (λ) measured in 50 mM HEPES buffer (pH 7.4, 0.2% DMSO) are 545 nm and 575 nm, respectively; extinction coefficient ε = 73,500 M-1cm-1; quantum yield φ = 0.19.

Please refer to this protocol for further information on how to use this product.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 4-(tert-Butoxycarbonyl)-1-(3'-(4-(tert-butoxycarbonyl)piperazin-1-yl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthen]-6'-yl)piperazine 1-oxide
CAS 2711762-70-6
Quantity 24 μg
Product Line Tocris
Target Ion Indicators
Molecular Formula C38H44N4O8
Purity 90%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.