missing translation for 'onlineSavingsMsg'
Learn More

Ferroin Indicator Solution, 0.025M, Honeywell Fluka™

Product Code. p-7120433 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass Bottle
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15645620 50 mL Glass Bottle 50mL
15635620 250 mL Glass Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15645620 Supplier Honeywell-Fluka Supplier No. 4627050ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Redox Indicator, 0.025 M

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
PubChem CID 84567
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 0°C
Boiling Point 100°C
Linear Formula [Fe(C12H8N2)3]SO4
UN Number NONH for all modes of transport
Formula Weight 692.52g/mol
Odor Odorless
Packaging Glass Bottle
Density 1.006g/mL at 20°C
Color Red
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Ferroin indicator solution
Show More Show Less
Product Identifier
  • Ferroin indicator solution
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Tris(1,10-phenanthroline-N1,N10)iron sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.