missing translation for 'onlineSavingsMsg'
Learn More

Ferrocene, TRC

Product Code. 30408548
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408548 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408548 Supplier Toronto Research Chemicals Supplier No. TRCF30759525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ferrocene
CAS 102-54-5
Molecular Formula 2 C5 H5 . Fe
Quantity 25 g
Synonym Ferrocene (8CI, 9CI, ACI), Bis(cyclopentadienyl)iron, Bis(η-cyclopentadienyl)iron, Bis(η5-cyclopentadienyl)iron, Catane TM, Di-2,4-cyclopentadien-1-yliron, Di-π-cyclopentadienyl iron, Dicyclopentadienyliron, Ferrotsen, Iron bis(cyclopentadienide), Iron dicyclopentadienyl, Iron, bis(η5-2,4-cyclopentadien-1-yl)-, NSC 2033, NSC 209798, NSC 44012, PLUTOcen F-C, PLUTOcen G
InChI Formula InChI=1S/2C5H5.Fe/c2*1-2-4-5-3-1;/h2*1-5H;
SMILES [Fe].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2
IUPAC Name cyclopentane;iron
Molecular Weight (g/mol) 186.03
Formula Weight 186.0132
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.