missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrocene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Melting Point | 173°C to 174°C |
Color | Yellow to Orange |
Boiling Point | 249°C |
Odor | Characteristic |
Quantity | 250 g |
UN Number | UN1325 |
Sensitivity | Air and moisture sensitive |
Merck Index | 14,4037 |
Solubility Information | Insoluble in water. Soluble in organic solvents. |
Formula Weight | 186.04 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'