missing translation for 'onlineSavingsMsg'
Learn More

Ferrocene, 98%

Product Code. p-4509564 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10224820

Brand: Thermo Scientific Acros 119141000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 102-54-5 | C10H10Fe | 186.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-54-5
Molecular Formula C10H10Fe
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001427
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym ferrocene, bis cyclopentadienyl iron
PubChem CID 25199998
IUPAC Name cyclopenta-1,3-diene;iron
SMILES [Fe].c1cccc1.c1cccc1

Specifications

Melting Point 173.0°C to 176.0°C
Color Brown-Orange
Boiling Point 249.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Merck Index 15,4064
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and benzene
Sublimation Point 0100.0°C
Formula Weight 186.04
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ferrocene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ferrocene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable solid Category 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.