missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Mouse anti-Human, Clone: FTH/2082, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Ferritin |
|---|---|
| Clone | FTH/2082 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Ferritin Monoclonal specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Ferritin | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 2495 | |
| IgG2c κ | |
| Protein A or G purified |
| FTH/2082 | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title