missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MIF2502 detects Ferritin in ELISA, IP, IF, and RIA applications and shows reactivity to human samples. The MIF2502 Immunogen is human liver ferritin. MIF2502 detects ferritin which has a predicted molecular weight of approximately 19kDa. MIF2502 was formerly sold as a Seradyn product.
Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. Ferritin is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in the light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. The ferritin gene has multiple pseudogenes, and ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. Ferritin is a holoenzyme shell with a molecular weight of about 450 kDa, and consists of 24 subunits of two types, H(heavy) and L(light), and capable of storing up to 4,500 atoms of ferric iron.
Specifications
Specifications
| Antigen | Ferritin |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 521 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FTL |
| Gene Accession No. | P02792, P02794 |
| Gene Alias | apoferritin; cell proliferation-inducing gene 15 protein; Ferritin H subunit; ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain, N-terminally processed; Ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3; Ferritin subunit H; ferritin, heavy polypeptide 1; ferritin, light polypeptide; FHC; frih; fril; FTH; Fth1; FTHL6; FTL; HFE5; H-ferritin; HFt; L apoferritin; LFTD; MFH; MGC71996; NBIA3; OK/SW-cl.84; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
