Learn More
Ferritin mitochondrial Antibody (AMB8LK), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43585
Product Code. 30279828
  • £516.00 / 0.10mg
Description
Ferritin mitochondrial Monoclonal antibody specifically detects Ferritin mitochondrial in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferritin mitochondrial
Monoclonal
1 mg/mL
Immunoprecipitation
EC 1.16.3.1, ferritin H subunit, ferritin mitochondrial, ferritin, mitochondrial, mitochondrial ferritin, MTF
The original antibody was generated by immunizing female Balb/c mice with ferritin extracted from human spleen cells. (Uniprot# Q8N4E7)
0.1 mg
Lipid and Metabolism
94033
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Immunoprecipitation
AMB8LK
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified