missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin mitochondrial Antibody (AMB8LK), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43586
This item is not returnable.
View return policy
Description
Ferritin mitochondrial Monoclonal antibody specifically detects Ferritin mitochondrial in Human samples. It is validated for Immunoprecipitation
Specifications
| Ferritin mitochondrial | |
| Monoclonal | |
| 1 mg/mL | |
| Immunoprecipitation | |
| EC 1.16.3.1, ferritin H subunit, ferritin mitochondrial, ferritin, mitochondrial, mitochondrial ferritin, MTF | |
| The original antibody was generated by immunizing female Balb/c mice with ferritin extracted from human spleen cells. (Uniprot# Q8N4E7) | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 94033 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunoprecipitation | |
| AMB8LK | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction