missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin light chain Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10727-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.Specifications
| Ferritin light chain | |
| Polyclonal | |
| Unconjugated | |
| FTL | |
| Ferritin L subunit, Ferritin light chain, ferritin, light polypeptide, FTL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14325, 2512, 29292 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02792, P02793, P29391 | |
| FTL, Ftl1 | |
| Ferritin light chain Fusion Protein Ag1149 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction