missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Light Chain Rabbit anti-Human, Clone: FTL/3872R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07805-100ug
This item is not returnable.
View return policy
Description
Ferritin Light Chain Monoclonal specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| Ferritin Light Chain | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3 | |
| Recombinant full-length human Ferritin Light Chain protein | |
| 100 μg | |
| Cancer | |
| 2512 | |
| Store at 4°C. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| FTL/3872R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction