missing translation for 'onlineSavingsMsg'
Learn More

Ferritin Light Chain Rabbit anti-Human, Clone: FTL/2338R, Novus Biologicals™

Rabbit Monoclonal Antibody

£412.00

Specifications

Antigen Ferritin Light Chain
Clone FTL/2338R
Concentration 0.2 mg/mL
Dilution Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18640907
View Documents
Novus Biologicals
NBP3-07804-100ug
100 μg
£412.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ferritin Light Chain Monoclonal specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

Ferritin Light Chain
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer
10 mM PBS with 0.05% BSA
2512
IgG
Protein A or G purified
FTL/2338R
Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3
Recombinant human Ferritin Light Chain protein fragment (around aa 38-165) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.