missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Light Chain Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142440
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes.Specifications
Ferritin Light Chain | |
Polyclonal | |
Biotin | |
FTL | |
ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril; Ftl; Ftl1; Ftl-1; L-ferritin; LFTD; NBIA3 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
14325, 2512, 29292 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P02792, P02793, P29391 | |
FTL, Ftl1 | |
Peptide with sequence C-GEYLFERLTLKHD. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |