missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin, Light Chain Mouse, Clone: FTL/1388, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2512-MSM3-P1
This item is not returnable.
View return policy
Specifications
| Ferritin, Light Chain | |
| Cytoplasmic | |
| FTL/1388 | |
| Unconjugated | |
| NP_000137 | |
| Mouse | |
| 19-25kDa | |
| Protein A/G Purified | |
| RUO | |
| 2512 | |
| IgG2a |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| FTL | |
| Recombinant human FTL protein fragment (aa 38-165) | |
| HepG2, HeLa, HL-60 or 293T cells. Pancreas, Liver, Cerebellum or Testis. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction