missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Light Chain Antibody (FTL/3872R), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08623AF647
This item is not returnable.
View return policy
Description
Ferritin Light Chain Monoclonal antibody specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Ferritin Light Chain | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3 | |
| Recombinant full-length human Ferritin Light Chain protein (Uniprot: P02792) | |
| 100 μg | |
| Cancer | |
| 2512 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| FTL/3872R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction