missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Light Chain Antibody (8E1E7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £463.00
Specifications
| Antigen | Ferritin Light Chain |
|---|---|
| Clone | 8E1E7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603756
|
Novus Biologicals
NBP2-37243-0.025ml |
0.025 mL |
£197.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179617
|
Novus Biologicals
NBP2-37243 |
0.1 mL |
£463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ferritin Light Chain Monoclonal specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Ferritin Light Chain | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3 | |
| FTL | |
| IgG1 | |
| Affinity Purified |
| 8E1E7 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Cancer | |
| PBS with 0.05% Sodium Azide | |
| 2512 | |
| Purified recombinant fragment of human FTL (AA: FULL(1-157)) expressed in E. Coli. | |
| Primary | |
| 20 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title