missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ferritin Heavy Chain Monoclonal Antibody (D4), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £385.00
Specifications
| Antigen | Ferritin Heavy Chain |
|---|---|
| Clone | D4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17644286
|
Invitrogen™
MA543830 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17634286
![]() |
Invitrogen™
MA543829 |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FTH1 is the heavy subunit of ferritin, the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in ferritin proteins are associated with several neurodegenerative diseases. This gene has multiple pseudogenes. Several alternatively spliced transcript variants have been observed, but their biological validity has not been determined.Specifications
| Ferritin Heavy Chain | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| apoferritin; cell proliferation-inducing gene 15 protein; Ferritin H subunit; ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain, N-terminally processed; Ferritin subunit H; ferritin, heavy polypeptide 1; FHC; frih; FTH; Fth1; FTHL6; HFE5; H-ferritin; HFt; MFH; MGC104426; OK/SW-cl.84; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15 | |
| FTH1 | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| D4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P02794, P19132 | |
| 2495, 25319 | |
| Met1-Ser183 (Accession P02794), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FTH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title