missing translation for 'onlineSavingsMsg'
Learn More

Ferritin Antibody, Novus Biologicals™

Product Code. 18379026 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379026 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379026 Supplier Novus Biologicals Supplier No. NBP314756

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Ferritin Polyclonal antibody specifically detects Ferritin in Human samples. It is validated for Immunoelectrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ferritin
Applications Immunoelectrophoresis
Classification Polyclonal
Conjugate Unconjugated
Formulation Phosphate Buffered Saline (PBS) containing with 0.09% Sodium Azide
Gene Alias apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Host Species Goat
Immunogen no immunogen data
Purification Method Multi-step
Quantity 2 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2495
Target Species Human
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.