missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Antibody (FTH/2082), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08613AF405
This item is not returnable.
View return policy
Description
Ferritin Monoclonal antibody specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Ferritin | |
| Monoclonal | |
| Alexa Fluor 405 | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary) (Uniprot: P02794) | |
| 100 μg | |
| Cancer, Cellular Markers, Lipid and Metabolism | |
| 2495 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| FTH/2082 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction