missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Antibody (321B-7 (05)), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62560AF532
This item is not returnable.
View return policy
Description
Ferritin Monoclonal antibody specifically detects Ferritin in Human samples. It is validated for ELISA
Specifications
| Ferritin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Ferritin from human liver. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Lipid and Metabolism | |
| 2495 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| 321B-7 (05) | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction