missing translation for 'onlineSavingsMsg'
Learn More

Ferric Phosphate, TRC

Product Code. 30135845
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135845 500 mg 500mg
30134641 1 g 1g
30012056 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135845 Supplier Toronto Research Chemicals Supplier No. TRCF307535500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10045-86-0
Molecular Formula Fe . O4 P
Molecular Weight (g/mol) 150.816
Synonym Iron Phosphate, Escargo!, Ferramol, Ferric Orthophosphate, Iron Orthophosphate, Iron Phosphate, Iron(III) Phosphate, Sluggo;
IUPAC Name iron(3+);phosphate
SMILES [Fe+3].[O-]P(=O)([O-])[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/Fe.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+3;/p-3
Formula Weight 150.888
Chemical Name or Material Ferric Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.