missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate Nonahydrate ACS MP Biomedicals

Product Code. 11430612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430612 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11430612 Supplier MP Biomedicals Supplier No. 0219404680
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS grade Ferric nitrate nonahydrate for research or further manufacturing use.

Specifications

CAS 7782-61-8
Physical Form Crystalline Powder
Quantity 100 g
Molecular Formula FeH18N3O18
MDL Number MFCD00149708
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name iron(3+) nonahydrate trinitrate
Molecular Weight (g/mol) 403.99
PubChem CID 16211566
Formula Weight 404.02
Percent Purity 99.2%
Grade ACS
Chemical Name or Material Ferric Nitrate, Nonahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.