missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate Dodecahydrate ACS MP Biomedicals

Product Code. 11490602 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490602 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11490602 Supplier MP Biomedicals Supplier No. 0219404425
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS grade reagent used as a mordant in dyeing, an analytical iron standard, and a catalyst.

Specifications

CAS 7783-83-7
Physical Form Crystalline Powder
Quantity 25 g
Density 1.71g/mL
Molecular Formula FeH28NO20S2
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
Molecular Weight (g/mol) 482.176
PubChem CID 197097
Formula Weight 482.2
Grade ACS
Chemical Name or Material Ferric Ammonium Sulfate, Dodecahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.