missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferredoxin Reductase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38055-100ul
This item is not returnable.
View return policy
Description
Ferredoxin Reductase Polyclonal antibody specifically detects Ferredoxin Reductase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Ferredoxin Reductase | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Adrenodoxin reductase, ADXREC 1.18.1.2, ferredoxin reductaseAR, Ferredoxin--NADP(+) reductase, NADPH:adrenodoxin oxidoreductase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 252-491 of human Ferredoxin Reductase (NP_077728.3).,, Sequence:, PILDPVDFLGLQDKIKEVPRPRKRLTELLLRTATEKPGPAEAARQASASRAWGLRFFRSPQQVLPSPDGRRAAGVRLAVTRLEGVDEATRAVPTGDMEDLPCGLVLSSIGYKSRPVDPSVPFDSKLGVIPNVEGRVMDVPGLYCSGWVKRGPTGVIATTMTDSFLTGQMLLQDLKAGLLPSGPRPGYAAIQALLSSRGVRPVSFSDWEKLDAEEVARGQGTGKPREKLVDPQEMLRLLGH | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2232 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction