missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FERMT3/URP2 Antibody (OTI3C7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72241B
This item is not returnable.
View return policy
Description
FERMT3/URP2 Monoclonal specifically detects FERMT3/URP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FERMT3/URP2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| FERMT3 | |
| Recombinant protein fragment corresponding to amino acids 259-528 of human FERMT3/URP2 (NP_113659) produced in E.coli. | |
| 0.1 mL | |
| 83706.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3C7 | |
| Western Blot, Immunohistochemistry | |
| fermitin family member 3, KIND3fermitin family homolog 3 (Drosophila), kindlin 3, Kindlin-3, MGC10966, MIG-2, MIG2BURP2SF, MIG2-like protein, UNC-112 related protein 2, UNC112C, Unc-112-related protein 2, URP2fermitin family homolog 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction