missing translation for 'onlineSavingsMsg'
Learn More

FERMT3/URP2 Antibody [DyLight 650], Novus Biologicals™

Product Code. 30732238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732238 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732238 Supplier Novus Biologicals Supplier No. AF7004W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

FERMT3/URP2 Polyclonal antibody specifically detects FERMT3/URP2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FERMT3/URP2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias fermitin family member 3, KIND3fermitin family homolog 3 (Drosophila), kindlin 3, Kindlin-3, MGC10966, MIG-2, MIG2BURP2SF, MIG2-like protein, UNC-112 related protein 2, UNC112C, Unc-112-related protein 2, URP2fermitin family homolog 3
Host Species Sheep
Immunogen E. coli-derived recombinant human FERMT3, Ala498-Phe667, Accession # Q86UX7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83706
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.