missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FERMT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FERMT1 |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16891745
|
Proteintech
22215-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16881745
|
Proteintech
22215-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Due to a concerted effort to identify biomarkers for lung and colon carcinomas by genome-wide transcriptional profiling, the identification and cloning of one such gene as well as two additional closely related genes was achieved. Due to the strong sequence homology to the C. elegans UNC-112 a novel gene was named URP1, for UNC-112 related protein. Another novel related gene, URP2 and the previously discovered MIG-2 gene was also identified. Transcriptional analysis shows that only URP1 is significantly differentially regulated, being over-expressed in 70% of the colon carcinomas and 60% of the lung carcinomas tested. Quantification of URP1 expression by qRT-PCR showed up-regulation of the gene by 60-fold in lung tumors and up to nearly 6-fold in colon tumors. Northern blot analysis of URP1 indicates that normal expression is restricted to neuromuscular tissues. In contrast, the expression of URP2 appears to be confined primarily to tissues of the immune system. URP1 has the potential to be one of the most important prognostic and diagnostic markers of colon or lung cancer.Specifications
| FERMT1 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| P59113, Q9BQL6 | |
| 241639, 55612 | |
| FERMT1 Fusion Protein Ag17543 | |
| Primary | |
| -20°C | |
| FERMT1 |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C20orf42, DTGCU2, Fermitin family homolog 1, FERMT1, KIND1, Kindlerin, Kindlin 1, Kindlin syndrome protein, Unc 112 related protein 1, UNC112A, URP1 | |
| FERMT1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title