missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ZipRuler Express DNA Ladder Set, ready-to-use
GREENER_CHOICE

Product Code. 11853993 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 50 μg
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11853993 2 x 50 μg Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11853993 Supplier Thermo Scientific™ Supplier No. SM1373

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Perform sizing and approximate quantification of wide range of double-stranded DNA for fast and long electrophoresis runs.

ZipRuler Express DNA Ladder Set contains two ladders: ZipRuler Express DNA Ladder 1 and ZipRuler Express DNA Ladder 2. Both are mixtures of chromatography-purified individual DNA fragments.The set is designed for fast and highly accurate sizing of a broad range (100-20,000 bp) of DNA fragments and their approximate quantification under a variety of electrophoresis conditions. The band pattern is not affected by the composition of electrophoresis buffer, voltage or gel percentage (see Table 1).

Highlights

  • Flexible -can be run in two lanes or combined in one lane
  • Composed of chromatography purified individual DNA fragments
  • Ready-to-use -premixed with a loading dye for direct loading and room temperature storage
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Orange G, Bromophenol Blue
Ready to Load Yes
Size Range 100 to 20000 bp
Gel Compatibility Agarose Gels
Quantity 2 x 50 μg
Product Line ZipRuler
Product Type DNA Ladder Set

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.