missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ VspI (AseI) (10 U/μL)

Product Code. 10329869 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,000 units
5,000 units
Unit Size:
1000 units
5000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10329869 1,000 units 1000 units
10639310 5,000 units 5000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10329869 Supplier Thermo Scientific™ Supplier No. ER0911

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Cut at AT^TAAT sites with VspI (AseI) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: AseI, PshBI).

Lambda DNA digested with VspI (AseI), 1% agarose, 17 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..A T^T AA T...3'

3'..T AA T^T A...5'

Conditions for 100% Activity

  • 1X Buffer O:50mM Tris-HCl (pH 7.5 at 37°C), 10 mM MgCl2, 100 mM NaCl and 0.1mg/ml BSA
  • Incubate at 37°C

Storage Buffer

  • VspI is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with VspI, more than 95% of DNA fragments can be ligated and recut

Methylation Effects

  • Dam: never overlaps – no effect
  • Dcm: never overlaps – no effect
  • CpG: never overlaps – no effect
  • EcoKI: never overlaps – no effect
  • EcoBI: may overlap – effect not determined

Digestion of Agarose-embedded DNA

  • Minimum 5 units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Compatible Ends

  • Csp6I, FspBI, NdeI, Tru1I
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme VspI (AseI)
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,000 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.