Learn More
Thermo Scientific™ Terminal Deoxynucleotidyl Transferase (20 U/μL)
Handla allt< titel> ProdukterBeskrivning
Terminal Deoxynucleotidyl Transferase (TdT), a template-independent DNA polymerase, catalyzes the repetitive addition of deoxyribonucleotides to the 3'-OH of oligodeoxyribonucleotides and single-stranded and double-stranded DNA. TdT requires an oligonucleotide of at least three nucleotides to serve as a primer. With RNA as template TdT shows variable performance which strongly depends upon the tertiary structure of acceptor RNA 3'-end and the nature of nucleotide. Generally, it is lower than using DNA as a template.
- Incorporates modified nucleotides (e.g., fluorescein-, biotin-, aminoallyl-labeled nucleotides) Source: E. coli cells carrying a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase
- The absence of endo-, exodeoxyribonucleases, phosphatases and ribonucleases confirmed by appropriate quality tests
Source:
- E.coli cells carrying a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase
- One unit of the enzyme catalyzes the incorporation of 1nmol of deoxythymidylate into a polynucleotide fraction (adsorbed on DE-81) in 60 min. at 37°C
- Enzyme activity is assayed in the following mixture:
- 200mM potassium cacodylate (pH 7.2), 1mM CoCl2, 0.01% (v/v) Triton X-100, 10μM oligo(dT)10, 1mM dTTP and 0.4MBq/ml [3H]-dTTP
- The enzyme is supplied in:100mM potassium acetate (pH 6.8), 2mM 2-mercaptoethanol, 0.01% (v/v) Triton X-100 and 50% (v/v) glycerol
- 1M potassium cacodylate, 125mM Tris, 0.05% (v/v) Triton X-100, 5mM CoCl2 (pH 7.2 at 25°C)
- Inhibitors: metal chelators, ammonium, chloride, iodide, phosphate ions
- Inactivated by heating at 70°C for 10 min. or by addition of EDTA
5X Reaction Buffer:
Inhibition and Inactivation:
Recommended for:
Production of synthetic homo- and heteropolymers (1); Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus (2, 3); Oligodeoxyribonucleotide and DNA labeling (2, 4-8); 5'-RACE (Rapid Amplification of cDNA Ends) (9); In situ localization of apoptosis (10)
Note:
Due to the presence of CoCl2 the TdT Reaction Buffer is incompatible with downstream applications. It is necessary to remove CoCl2 from the reaction mixture by spin column or phenol/chloroform extraction and subsequent ethanol precipitation.
Specifikationer
Specifikationer
| Concentration | 20 U/μL |
| Enzyme | Terminal Deoxynucleotidyl Transferase |
| Compatible Buffer | Storage Buffer, 5X Reaction Buffer |
| Quantity | 500 units |
| Product Type | TdT |
For Research Use Only. Not for use in diagnostic procedures.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.