missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Terminal Deoxynucleotidyl Transferase (20 U/μL)

Product Code. 10745661 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2,500 units
500 units
Unit Size:
2500 units
500 units
This item is not returnable. View return policy

Product Code. 10745661

Brand: Thermo Scientific™ EP0162

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Terminal Deoxynucleotidyl Transferase (TdT) catalyzes the template-independent addition of deoxyribonucleotides to the 3'-OH terminus of DNA molecules.

Terminal Deoxynucleotidyl Transferase (TdT), a template-independent DNA polymerase, catalyzes the repetitive addition of deoxyribonucleotides to the 3'-OH of oligodeoxyribonucleotides and single-stranded and double-stranded DNA. TdT requires an oligonucleotide of at least three nucleotides to serve as a primer. With RNA as template TdT shows variable performance which strongly depends upon the tertiary structure of acceptor RNA 3'-end and the nature of nucleotide. Generally, it is lower than using DNA as a template.

  • Incorporates modified nucleotides (e.g., fluorescein-, biotin-, aminoallyl-labeled nucleotides) Source: E. coli cells carrying a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase
Quality Control:
  • The absence of endo-, exodeoxyribonucleases, phosphatases and ribonucleases confirmed by appropriate quality tests

Source:

  • E.coli cells carrying a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase
Definition of Activity Unit:
  • One unit of the enzyme catalyzes the incorporation of 1nmol of deoxythymidylate into a polynucleotide fraction (adsorbed on DE-81) in 60 min. at 37°C
  • Enzyme activity is assayed in the following mixture:
    • 200mM potassium cacodylate (pH 7.2), 1mM CoCl2, 0.01% (v/v) Triton X-100, 10μM oligo(dT)10, 1mM dTTP and 0.4MBq/ml [3H]-dTTP
    Storage Buffer:
    • The enzyme is supplied in:100mM potassium acetate (pH 6.8), 2mM 2-mercaptoethanol, 0.01% (v/v) Triton X-100 and 50% (v/v) glycerol

    5X Reaction Buffer:

    • 1M potassium cacodylate, 125mM Tris, 0.05% (v/v) Triton X-100, 5mM CoCl2 (pH 7.2 at 25°C)

    Inhibition and Inactivation:

    • Inhibitors: metal chelators, ammonium, chloride, iodide, phosphate ions
    • Inactivated by heating at 70°C for 10 min. or by addition of EDTA

    Recommended for:

    Production of synthetic homo- and heteropolymers (1); Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus (2, 3); Oligodeoxyribonucleotide and DNA labeling (2, 4-8); 5'-RACE (Rapid Amplification of cDNA Ends) (9); In situ localization of apoptosis (10)

    Note:

    Due to the presence of CoCl2 the TdT Reaction Buffer is incompatible with downstream applications. It is necessary to remove CoCl2 from the reaction mixture by spin column or phenol/chloroform extraction and subsequent ethanol precipitation.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 U/μL
Enzyme Terminal Deoxynucleotidyl Transferase
Compatible Buffer Storage Buffer, 5X Reaction Buffer
Quantity 2,500 units
Product Type TdT

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.