missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T3 RNA Polymerase

Product Code. 10849020 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10,000 U
Unit Size:
10000 units
This item is not returnable. View return policy

Product Code. 10849020

Brand: Thermo Scientific™ EP0102

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ Fermentas T3 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.

Thermo Scientific™ Fermentas Bacteriophage T3 RNA polymerase is DNA-dependent RNA polymerase with strict specificity for their respective double-stranded promoters.

It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from its promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)

Recommended for:

Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.

Note:

Consensus promoter sequences: AATTAACCCTCACTAAA G GGAGA–T3 TAATACGACTCACTATA G GGAGA–T7 ATTTAGGTGACACTATA G AAGNG–SP6 The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively (9).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20U/µL
Description T3 RNA Polymerase
For Use With (Application) Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
Includes 1mL of 5X Transcription Buffer
Quantity 10,000 U
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.