missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T3 promoter Sequencing Primer, 24-mer

Product Code. 10121330 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 μM, 42 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121330 10 μM, 42 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10121330 Supplier Thermo Scientific™ Supplier No. SO120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends.

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends. The primers are complementary to the T3 RNA Polymerase promoter region and are supplied as 10 µM aqueous solutions.

Applications
• Sequencing of DNA fragments located downstream from the T3 RNA polymerase promoter sequence in common cloning vectors, such as pTZ19R (Cat. No. SD0141), pTZ57R, and pBluescript II

Promoter Sequence 5'-d (GCGCGAAATTAACCCTCACTAAAG)-3'

Related products
SP6 promoter Sequencing Primer, 18-mer (SO116)
T3 promoter Sequencing Primer, 17-mer (SO119)
SP6 promoter Sequencing Primer, 24-mer (SO117)
T7 promoter Sequencing Primer, 20-mer (SO118)

Notes
• Primers cannot be used for certain plasmids that contain truncated, but still fully functional promoters
• Primers are not phosphorylated

TRUSTED_SUSTAINABILITY

Specifications

Promoter SP6, T3, T7
Product Type Sequencing Primer
Content And Storage T3 promoter Sequencing Primer, 24-mer, 10 μM (42 μL)

Store at –20°C.
Shipping Condition Dry Ice
Concentration 10 μM
Primer T3
Quantity 10 μM, 42 μL
For Use With (Application) Sequencing
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.