missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ScaI (10 U/μL)

Product Code. 10600081 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1,000 units
5,000 units
Unit Size:
1000 units
5000 units
This item is not returnable. View return policy

Product Code. 10600081

Brand: Thermo Scientific™ ER0431

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The ScaI restriction enzyme recognizes AGT^ACT sites and cuts best at 37°C in its own unique buffer (Isoschizomers: AssI, BmcAI, ZrmI).

Lambda DNA digested with ScaI, 0.7% agarose, 5 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...A G T▵A C T...3'

3'...T C A▵T G A...5'

Conditions for 100% Activity

  • 1X Buffer ScaI:10mM Bis-Tris Propane-HCl (pH6.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer

  • ScaI is supplied in:10mM Tris-HCl (pH7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with ScaI, more than 80% of the DNA fragments can be ligated and more than 90% of these can be recut

Methylation Effects

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: never overlaps — no effect
  • EcoKI: never overlaps — no effect
  • EcoBI: may overlap — blocked

Digestion of Agarose-embedded DNA

  • Minimum 20units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours

Note:

Conditions of low salt, high enzyme concentration, glycerol concentration > 5% or pH> 8.0 may result in star activity. Supercoiled plasmids may require up to 20-fold more ScaI for complete digestion than linear DNA.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Sca I
Compatible Buffer Unique Buffer (10X Buffer ScaI)
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,000 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.