Learn More
Thermo Scientific™ SatI (Fnu4HI) (10 U/μL)
Shop All Thermo Scientific ProductsDescription
Lambda DNA digested with SatI (Fnu4HI), 1.4% agarose, 380 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
3'...C G N▵C G...5'
Conditions for 100% Activity
- 1X Buffer G:10mM Tris-HCl (pH7.5 at 37°C), 10mM MgCl2, 50mM NaCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer
- SatI is supplied in:10mM Tris-HCl (pH7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage
- After 50-fold overdigestion with SatI, more than 60% of the DNA fragments can be ligated in a reaction mixture containing 10-30U of T4 DNA Ligase/1μg of fragments and 10% PEG
- More than 90% of these can be recut
Methylation Effects
- Dam: never overlaps — no effect
- Dcm: never overlaps — no effect
- CpG: may overlap — blocked
- EcoKI: never overlaps — no effect
- EcoBI: never overlaps — no effect
Compatible Ends
- GC^SGC — BcnI, Bme1390I
- GC^WGC — MvaI, Bme1390I
Note:
At least two copies of SatI (Fnu4HI) recognition site are required for efficient cleavage.
Specifications
Specifications
| Concentration | 10 U/μL |
| Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
| Enzyme | SatI (Fnu4HI) |
| Compatible Buffer | 10X Buffer G |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 200 units |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.