missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ RNase H (5 U/μL)
Ribonuclease H (RNase H) is an endoribonuclease that specifically degrades the RNA strand in RNA-DNA hybrids.
£163.00 - £660.00
Specifications
Concentration | 5 U/μL |
---|---|
Enzyme | RNase |
Compatible Buffer | 10X Reaction Buffer |
Product Type | RNase H |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10334860
|
Thermo Scientific™
EN0201 |
100 U |
£163.00
100 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10519310
|
Thermo Scientific™
EN0202 |
500 U |
£660.00
500 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ribonuclease H (Rnase H) specifically degrades the RNA strand in RNA-DNA hybrids. It does not hydrolyze the phosphodiester bonds within single-stranded and double-stranded DNA and RNA.
Applications- Removal of mRNA prior to synthesis of second strand cDNA
- RT-PCR and qRT-PCR: removal of RNA after first strand cDNA synthesis
- Removal of the poly(A) sequences of mRNA after hybridization with oligo(dT)
- Site-specific cleavage of RNA
- Studies of in vitro polyadenylation reaction products
Specifications
5 U/μL | |
RNase | |
10X Reaction Buffer | |
RNase H |
For Research Use Only. Not for use in diagnostic procedures.